tert-Butyl 4-(2-aminoacetyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A224690-1G A224690-250MG
77013-356EA 116.7 USD
77013-356 77013-358
tert-Butyl 4-(2-aminoacetyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(2-aminoacetyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁N₃O₃
MW: 243.30 g/mol
MDL Number: MFCD10011045
CAS Number: 352359-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR