Ethyl-1-acetylcyclopropane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-acetylcyclopropanecarboxylate

A224600-100G A224600-250MG A224600-25G A224600-5G A224600-1G
77030-904EA 580 USD
77030-904 77872-144 77872-145 77872-146 77872-143
Ethyl-1-acetylcyclopropane-1-carboxylate 97%
Ethyl-1-acetylcyclopropane-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MDL Number: MFCD00480999
CAS Number: 32933-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR