(3S,4S)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

A224380-100MG A224380-250MG
77013-350EA 89.12 USD
77013-350 77612-180
(3S,4S)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate 98%
(3S,4S)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18632749
CAS Number: 1174020-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR