(R)-2-(Pyrrolidin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A224286-100MG A224286-1G A224286-250MG
76987-124EA 26.53 USD
76987-124 76987-126 76987-128
(R)-2-(Pyrrolidin-2-yl)acetic acid 95%
(R)-2-(Pyrrolidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD11505916
CAS Number: 61350-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR