1-Cbz-3-Cyanoazetidine 95%

Supplier: Ambeed
Warning

A223949-100MG A223949-1G A223949-250MG A223949-5G
77030-886EA 6.37 USD
77030-886 77030-888 77030-890 77030-892
1-Cbz-3-Cyanoazetidine 95%
1-Cbz-3-Cyanoazetidine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
MDL Number: MFCD11110707
CAS Number: 288851-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR