[3,4-Bis(acetyloxy)-5-(2,6-dichloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,6-dichloro-9H-purin-9-yl)tetrahydrofuran-3,4-diyl diacetate

A223907-100MG A223907-1G A223907-250MG A223907-25G A223907-5G
77000-004EA 8 USD
77000-004 77000-006 77000-008 77000-010 77000-012
[3,4-Bis(acetyloxy)-5-(2,6-dichloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate 97%
[3,4-Bis(acetyloxy)-5-(2,6-dichloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₆Cl₂N₄O₇
MW: 447.227 g/mol
MDL Number: MFCD02093101
CAS Number: 3056-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR