2-Chloro-4-fluorobenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-fluorobenzo[d]thiazole

A223706-1G A223706-250MG A223706-5G
76999-998EA 54.11 USD
76999-998 77000-000 77000-002
2-Chloro-4-fluorobenzothiazole 98%
2-Chloro-4-fluorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₃ClFNS
MW: 187.62 g/mol
MDL Number: MFCD04448820
CAS Number: 182344-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR