3-(3-Methoxyphenyl)pyrrolidine oxalate(2:1) 95%

Supplier: Ambeed
Warning

A223556-1G A223556-250MG
77013-338EA 130.5 USD
77013-338 77013-340
3-(3-Methoxyphenyl)pyrrolidine oxalate(2:1) 95%
3-(3-Methoxyphenyl)pyrrolidine oxalate(2:1)

Precaución: for Research Use Only

Formula: C₂₄H₃₂N₂O₆
MW: 444.52 g/mol
MDL Number: MFCD28016703
CAS Number: 1610028-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR