Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2,1,3-Benzothiadiazole-5-boronic acid, pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole, Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester

A223550-100MG A223550-1G A223550-250MG
77013-332EA 26 USD
77013-332 77013-334 77013-336
Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester 95%
Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂S
MDL Number: MFCD11867874
CAS Number: 1168135-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR