5-Methylthiazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A223442-100MG A223442-25G
77013-322EA 10.61 USD
77013-322 77013-324
5-Methylthiazole-4-carboxylic acid 98%
5-Methylthiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C5H5NO2S
MW: 143.1600 g/mol
MDL Number: MFCD06738813
CAS Number: 120237-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR