Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,β-diaminopropionic acid 98%

Supplier: Ambeed
Warning

A223439-100MG A223439-1G A223439-250MG A223439-5G
77013-314EA 17 USD
77013-314 77013-316 77013-318 77013-320
Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,β-diaminopropionic acid 98%
Fmoc-(N-beta-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-alpha,beta-diaminopropionic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₂O₆
MDL Number: MFCD01631974
CAS Number: 247127-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR