N-(6-Bromoimidazo[1,2-a]pyridin-2-yl)-2,2,2-trifluoroacetamide 97%

Supplier: Ambeed
Warning

A223288-100MG A223288-1G A223288-250MG
76987-100EA 33.95 USD
76987-100 76987-102 76987-104
N-(6-Bromoimidazo[1,2-a]pyridin-2-yl)-2,2,2-trifluoroacetamide 97%
N-(6-Bromoimidazo[1,2-a]pyridin-2-yl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₉H₅BrF₃N₃O
MDL Number: MFCD11976190
CAS Number: 504413-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR