4-Iodo-3-(trifluoromethyl)benzoic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 5-Carboxy-2-iodobenzotrifluoride

A223225-1G A223225-5G A223225-250MG
76987-096EA 11 USD
76987-096 76987-098 77683-644
4-Iodo-3-(trifluoromethyl)benzoic acid 97%, Ambeed.Inc
4-Iodo-3-(trifluoromethyl)benzoic acid

Precaución: For research use only.

Formula: C₈H₄F₃IO₂
MDL Number: MFCD09258947
CAS Number: 914636-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR