Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Supplier: Ambeed
|
Warning
|
A223116-5G
A223116-1G
76987-094EA
333.13
USD
76987-094
77683-640
Methyl 4-bromo-2-chloro-6-fluorobenzoate 97%
Methyl 4-bromo-2-chloro-6-fluorobenzoate
Precaución: For research use only.
Formula:
C₈H₅BrClFO₂ MW: 267.48 g/mol |
MDL Number:
MFCD22574196 CAS Number: 1321613-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...