2,2,2,3',5'-Pentafluoroacetophenone 97%

Supplier: Ambeed
Warning

A222968-10G A222968-25G A222968-1G
76987-090EA 40 USD
76987-090 76987-092 77890-315
2,2,2,3',5'-Pentafluoroacetophenone 97%
2,2,2,3',5'-Pentafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃F₅O
MDL Number: MFCD01319993
CAS Number: 845823-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR