5-Methoxyindole-3-acetonitrile 98%

Supplier: Ambeed
Warning

A222749-1G A222749-250MG A222749-25G A222749-5G
77030-870EA 22 USD
77030-870 77030-872 77030-874 77030-876
5-Methoxyindole-3-acetonitrile 98%
5-Methoxyindole-3-acetonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O
MDL Number: MFCD02094163
CAS Number: 2436-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR