Dibenzyl chloromethyl phosphate 97%

Supplier: Ambeed
Warning

Synonyms: Dibenzyl (chloromethyl) phosphate

A222660-250MG A222660-5G
76987-086EA 24.41 USD
76987-086 76987-088
Dibenzyl chloromethyl phosphate 97%
Dibenzyl chloromethyl phosphate

Precaución: for Research Use Only

Formula: C₁₅H₁₆ClO₄P
MW: 326.71 g/mol
MDL Number: MFCD16652586
CAS Number: 258516-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR