2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole 95%

Supplier: Ambeed
Warning

A222644-100MG A222644-10G A222644-1G A222644-250MG A222644-5G
76999-974EA 28 USD
76999-974 76999-976 76999-978 76999-980 76999-982
2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole 95%
2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole

Precaución: for Research Use Only

Formula: C₈H₆N₄O₂S
MDL Number: MFCD00453655
CAS Number: 833-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR