5-Amino-4,6-dichloro-2-(propylthio)pyrimidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,6-Dichloro-2-(propylthio)pyrimidin-5-amine
A222402-100G
A222402-10G
A222402-25G
A222402-500G
A222402-5G
77030-860EA
117.76
USD
77030-860
77030-862
77030-864
77030-866
77030-868
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine 98%
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine
Precaución: for Research Use Only
Formula:
C₇H₉Cl₂N₃S MW: 238.13 g/mol |
MDL Number:
MFCD16875503 CAS Number: 145783-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...