Melamine 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine

A222399-100G A222399-25G A222399-500G A222399-1000G
77013-292EA 8.49 USD
77013-292 77013-294 77013-296 77568-146
Melamine 98%
Melamine

Precaución: For research use only.

Formula: C₃H₆N₆
MW: 126.12 g/mol
MDL Number: MFCD00006055
CAS Number: 108-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR