5-Hydroxynorbornane-2,6-lactone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one

A222386-1G A222386-5G
77030-856EA 5.31 USD
77030-856 77030-858
5-Hydroxynorbornane-2,6-lactone 98%
5-Hydroxynorbornane-2,6-lactone

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
MDL Number: MFCD00988184
CAS Number: 92343-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR