5-Benzyl-4,5,6,7-tetrahydro-5-azaindole 95%

Supplier: Ambeed
Warning

A222221-1G A222221-250MG A222221-5G
77030-850EA 331 USD
77030-850 77030-852 77030-854
5-Benzyl-4,5,6,7-tetrahydro-5-azaindole 95%
5-Benzyl-4,5,6,7-tetrahydro-5-azaindole

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂
MDL Number: MFCD02320628
CAS Number: 272442-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR