4-Methoxy-1-naphthaleneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Methoxynaphthalen-1-yl)boronic acid, 4-Methoxynaphthalene-1-boronic acid

A222212-1G A222212-5G A222212-250MG
76987-084EA 24 USD
76987-084 77890-314 77890-313
4-Methoxy-1-naphthaleneboronic acid 98%
4-Methoxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
MDL Number: MFCD03452757
CAS Number: 219834-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR