1H-Indazole-6-boronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
A222204-100MG
A222204-10G
A222204-1G
A222204-250MG
77030-846EA
10
USD
77030-846
77030-848
77600-778
77872-136
1H-Indazole-6-boronic acid pinacol ester 98%
1H-Indazole-6-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BN₂O₂ MW: 244.0973 g/mol |
MDL Number:
MFCD07368067 CAS Number: 937049-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...