(4-(Thiazol-2-yl)phenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A222052-250MG A222052-5G
77013-288EA 64.72 USD
77013-288 77013-290
(4-(Thiazol-2-yl)phenyl)methanamine hydrochloride 97%
(4-(Thiazol-2-yl)phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂S
MW: 226.73 g/mol
MDL Number: MFCD14525839
CAS Number: 1187451-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR