2-[4-(2-Pyrimidyl)-1-piperazinyl]acetic Acid 95%

Supplier: Ambeed
Warning

A221850-5G A221850-250MG
77030-844EA 159 USD
77030-844 77872-132
2-[4-(2-Pyrimidyl)-1-piperazinyl]acetic Acid 95%
2-[4-(2-Pyrimidyl)-1-piperazinyl]acetic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₄O₂
MDL Number: MFCD09711499
CAS Number: 244255-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR