2-Cyano-5-hydroxymethylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Hydroxymethyl)picolinonitrile, 5-(Hydroxymethyl)pyridine-2-carbonitrile

A221640-1G A221640-250MG A221640-5G
77013-274EA 140.04 USD
77013-274 77013-276 77013-278
2-Cyano-5-hydroxymethylpyridine 95%
2-Cyano-5-hydroxymethylpyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD09029606
CAS Number: 58553-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR