4-Bromo-2-chloro-5-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A221537-100G A221537-10G A221537-1G A221537-250MG A221537-25G
76999-940EA 327.82 USD
76999-940 76999-942 76999-944 76999-946 76999-948
4-Bromo-2-chloro-5-fluorobenzoic acid 97%
4-Bromo-2-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MDL Number: MFCD00236589
CAS Number: 177480-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR