2-Amino-6-(trifluoromethoxy)benzothiazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

A221527-100MG A221527-1G A221527-25G A221527-5G
76999-932EA 5 USD
76999-932 76999-934 76999-936 76999-938
2-Amino-6-(trifluoromethoxy)benzothiazole 97%
2-Amino-6-(trifluoromethoxy)benzothiazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂OS
MDL Number: MFCD00210213
CAS Number: 1744-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR