1-(2-Chloropyridin-3-yl)ethanol 96%

Supplier: Ambeed
Warning

A221392-100MG A221392-250MG
76987-072EA 14.86 USD
76987-072 76987-074
1-(2-Chloropyridin-3-yl)ethanol 96%
1-(2-Chloropyridin-3-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD16037360
CAS Number: 131674-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR