2-Amino-5-chloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A221308-100G A221308-10G A221308-5G A221308-25G
76999-924EA 63 USD
76999-924 76999-926 76999-928 77881-550
2-Amino-5-chloro-3-methylpyridine 98%
2-Amino-5-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD00129030
CAS Number: 20712-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR