(4-Methoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A221202-1000G A221202-100G A221202-10G A221202-25G A221202-500G
77030-814EA 99 USD
77030-814 77030-816 77030-818 77030-820 77030-822
(4-Methoxyphenyl)acetic acid 98%
(4-Methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(OCH₃))CH₂COOH
MDL Number: MFCD00004345
CAS Number: 104-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR