(R)-tert-Butyl (2-oxopiperidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A221198-100MG A221198-250MG A221198-1G
77030-810EA 24 USD
77030-810 77030-812 77872-127
(R)-tert-Butyl (2-oxopiperidin-3-yl)carbamate 95%
(R)-tert-Butyl (2-oxopiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD09831897
CAS Number: 221874-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR