4-Chloro-5-fluoro-2-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A221156-10G A221156-1G A221156-250MG A221156-25G A221156-5G
76987-062EA 548.49 USD
76987-062 76987-064 76987-066 76987-068 76987-070
4-Chloro-5-fluoro-2-methoxypyrimidine 97%
4-Chloro-5-fluoro-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂O
MW: 162.549 g/mol
MDL Number: MFCD09834963
CAS Number: 1801-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR