Methyl 5-bromo-2-ethylbenzoate 98%

Supplier: Ambeed
Warning

A221080-100MG A221080-1G A221080-250MG
76987-054EA 6 USD
76987-054 76987-056 76987-058
Methyl 5-bromo-2-ethylbenzoate 98%
Methyl 5-bromo-2-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD11112975
CAS Number: 439937-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR