4-Bromo-5-methoxybenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A220983-100MG A220983-250MG
76987-050EA 50 USD
76987-050 76987-052
4-Bromo-5-methoxybenzene-1,2-diamine 95%
4-Bromo-5-methoxybenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MDL Number: MFCD21648471
CAS Number: 108447-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR