3-(4-Methylpiperazin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A220934-1G A220934-250MG
76999-910EA 39.26 USD
76999-910 76999-912
3-(4-Methylpiperazin-1-yl)benzoic acid 95%
3-(4-Methylpiperazin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD06742238
CAS Number: 215309-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR