3,5-Difluoro-4-(trifluoromethyl)phenol 97%

Supplier: Ambeed
Warning

A220837-1G A220837-250MG
77013-256EA 370 USD
77013-256 77013-258
3,5-Difluoro-4-(trifluoromethyl)phenol 97%
3,5-Difluoro-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃F₅O
MDL Number: MFCD19441183
CAS Number: 116640-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR