2-Chloro-3-iodo-5-nitropyridine 95%

Supplier: Ambeed
Danger

A220691-1G A220691-5G
77013-254EA 31 USD
77013-254 77612-150
2-Chloro-3-iodo-5-nitropyridine 95%
2-Chloro-3-iodo-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClIN₂O₂
MDL Number: MFCD12028701
CAS Number: 25391-60-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR