4-(Chlorosulfonyl)phenyl pivalate 95%

Supplier: Ambeed
Danger

Synonyms: (4-Chlorosulphonylphenyl) 2,2-dimethylpropanoate

A220665-100MG A220665-1G
77030-806EA 52 USD
77030-806 77030-808
4-(Chlorosulfonyl)phenyl pivalate 95%
4-(Chlorosulfonyl)phenyl pivalate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₄S
MDL Number: MFCD08274725
CAS Number: 150374-99-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR