1-Bromo-3-methylbutan-2-one 95%

Supplier: Ambeed
Danger

A220648-100MG A220648-1G A220648-10G A220648-5G
77030-800EA 6.37 USD
77030-800 77030-802 77550-432 77667-836
1-Bromo-3-methylbutan-2-one 95%
1-Bromo-3-methylbutan-2-one

Precaución: for Research Use Only

Formula: C₅H₉BrO
MW: 165.0284 g/mol
MDL Number: MFCD00015716
CAS Number: 19967-55-6
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR