2-Mercaptobenzothiazole 98%

Supplier: Ambeed
Danger

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

A220622-1000G A220622-100G A220622-25G A220622-500G
76999-902EA 32.89 USD
76999-902 76999-904 76999-906 76999-908
2-Mercaptobenzothiazole 98%
2-Mercaptobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅NS₂
MW: 167.26 g/mol
MDL Number: MFCD00005781
CAS Number: 149-30-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR