5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A220416-100MG A220416-1G A220416-250MG
76987-036EA 7 USD
76987-036 76987-038 76987-040
5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD12405111
CAS Number: 507462-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR