4-Bromo-3,5-dimethylisoxazole 98%

Supplier: Ambeed
Warning

A220057-100G A220057-10G A220057-1G A220057-25G A220057-5G
77013-228EA 65 USD
77013-228 77013-230 77013-232 77013-234 77013-236
4-Bromo-3,5-dimethylisoxazole 98%
4-Bromo-3,5-dimethylisoxazole

Precaución: for Research Use Only

Formula: C₅H₆BrNO
MDL Number: MFCD00068187
CAS Number: 10558-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR