4-Amino-3-cyanobenzoic acid 95%

Supplier: Ambeed
Warning

A219875-100MG A219875-1G A219875-250MG A219875-5G
77013-222EA 33.95 USD
77013-222 77013-224 77013-226 77683-634
4-Amino-3-cyanobenzoic acid 95%
4-Amino-3-cyanobenzoic acid

Precaución: For research use only.

Formula: C₈H₆N₂O₂
MW: 162.145 g/mol
MDL Number: MFCD09745178
CAS Number: 74192-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR