1,3-Oxazolo[4,5-b]pyridine-2-thiol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Thio-1,3-oxazolo[4,5-b]pyridine, Oxazolo[4,5-b]pyridine-2-thiol

A219761-1G A219761-250MG
77030-764EA 50.93 USD
77030-764 77030-766
1,3-Oxazolo[4,5-b]pyridine-2-thiol 98%
1,3-Oxazolo[4,5-b]pyridine-2-thiol

Precaución: for Research Use Only

Formula: C₆H₄N₂OS
MW: 152.174 g/mol
MDL Number: MFCD00973914
CAS Number: 211949-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR