Telmisartan 98%

Supplier: Ambeed
Warning

A219723-100G A219723-100MG A219723-10G A219723-1G A219723-25G A219723-500G A219723-5G
76999-878EA 94 USD
76999-878 76999-880 76999-882 76999-884 76999-886 76999-888 76999-890
Telmisartan 98%
Telmisartan

Precaución: for Research Use Only

Formula: C₃₃H₃₀N₄O₂
MDL Number: MFCD00918125
CAS Number: 144701-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR