3-Bromo-5-fluoropyridin-4-amine 95%
Supplier: Ambeed
|
Warning
|
A219641-1G
A219641-250MG
A219641-5G
A219641-10G
77030-760EA
49
USD
77030-760
77030-762
77568-124
77872-113
3-Bromo-5-fluoropyridin-4-amine 95%
3-Bromo-5-fluoropyridin-4-amine
Precaución: For research use only.
Formula:
C5H4BrFN2 |
MDL Number:
MFCD13185327 CAS Number: 1214326-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...