Guaiacylglycerol-β-guaiacyl ether 98%

Supplier: Ambeed
Danger

A219596-1G A219596-250MG
77030-756EA 495 USD
77030-756 77030-758
Guaiacylglycerol-β-guaiacyl ether 98%
Guaiacylglycerol-beta-guaiacyl ether

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₆
MDL Number: MFCD00060144
CAS Number: 7382-59-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR