Methyl 2-amino-6-methylisonicotinate 95%

Supplier: Ambeed
Warning

A219470-1G A219470-250MG
77013-218EA 643.97 USD
77013-218 77013-220
Methyl 2-amino-6-methylisonicotinate 95%
Methyl 2-amino-6-methylisonicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.17 g/mol
MDL Number: MFCD16611578
CAS Number: 1029128-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR